Tisk…
Anti-SMC1A Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB137707-100EA 25740 CZK
ABCAAB137707-100
Anti-SMC1A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SMC1A (structural maintenance of chromosomes 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human