Tisk…

Anti-AIFM1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137725-100EA 26320 CZK
ABCAAB137725-100
Anti-AIFM1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AIFM1 (apoptosis-inducing factor, mitochondrion-associated, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR