Tisk…

Anti-AP2M1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137727-100EA 25800 CZK
ABCAAB137727-100
Anti-AP2M1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AP2M1 (adaptor-related protein complex 2, mu 1 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR