Tisk…

Anti-CYC1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137757-100EA 25590 CZK
ABCAAB137757-100
Anti-CYC1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CYC1 (cytochrome c-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR