Tisk…
Anti-TRAF5 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB137763-100EA 25590 CZK
ABCAAB137763-100
Anti-TRAF5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRAF5 (TNF receptor-associated factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human