Tisk…

Anti-FBXW11 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137835-100EA 24730 CZK
ABCAAB137835-100
Anti-FBXW11 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FBXW11 (F-box and WD repeat domain containing 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR