Tisk…

Anti-HA tag Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137838-100EA 26510 CZK
ABCAAB137838-100
Anti-HA tag Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR