Tisk…

Anti-14-3-3 epsilon Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137862-100EA 26320 CZK
ABCAAB137862-100
Anti-14-3-3 epsilon Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: 14-3-3 epsilon (14-3-3 epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR