Tisk…

Anti-POLR3A Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB138023-100EA 25740 CZK
ABCAAB138023-100
Anti-POLR3A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: POLR3A (polymerase (RNA) III (DNA directed) polypeptide A, 155kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR