Tisk…
Anti-RPL36 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB138032-100EA 25590 CZK
ABCAAB138032-100
Anti-RPL36 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RPL36 (ribosomal protein L36)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human