Tisk…

Anti-MRPL14 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB138281-100EA 24420 CZK
ABCAAB138281-100
Anti-MRPL14 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MRPL14 (mitochondrial ribosomal protein L14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR