Tisk…
Anti-GGH Rabbit Monoclonal Antibody
Dodavatel: Abcam
ABCAAB138495-100EA 25740 CZK
ABCAAB138495-100
Anti-GGH Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: GGH
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat