Tisk…
Anti-HOXD10 Rabbit Monoclonal Antibody
Dodavatel: Abcam
ABCAAB138508-100EA 25460 CZK
ABCAAB138508-100
Anti-HOXD10 Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: HOXD10
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat