Tisk…

Anti-PEA15 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB138647-100EA 26080 CZK
ABCAAB138647-100
Anti-PEA15 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PEA15 (phosphoprotein enriched in astrocytes 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR