Tisk…

Anti-BAX Rabbit Monoclonal Antibody (FITC (Fluorescein)) [clone: T22-A]

Dodavatel: Abcam
ABCAAB139543-50EA 25560 CZK
ABCAAB139543-50
Anti-BAX Rabbit Monoclonal Antibody (FITC (Fluorescein)) [clone: T22-A]
Protilátky
Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: monoclonal
Clone: T22-A
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR