Tisk…

Anti-VPS13B Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB139814-100EA 24420 CZK
ABCAAB139814-100
Anti-VPS13B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: VPS13B (vacuolar protein sorting 13 homolog B (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR