Tisk…
Anti-CARD10 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB140204-100EA 25740 CZK
ABCAAB140204-100
Anti-CARD10 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CARD10 (caspase recruitment domain family, member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human