Tisk…

Anti-TUBGCP2 Mouse Monoclonal Antibody [clone: GCP2-01]

Dodavatel: Abcam
ABCAAB140225-100EA 24420 CZK
ABCAAB140225-100
Anti-TUBGCP2 Mouse Monoclonal Antibody [clone: GCP2-01]
Protilátky
Type: Primary
Antigen: TUBGCP2 (tubulin, gamma complex associated protein 2)
Clonality: monoclonal
Clone: GCP2-01
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Pig, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR