Tisk…
Anti-MelanA Mouse Monoclonal Antibody [clone: 3E2]
Dodavatel: Abcam
ABCAAB140503-100EA 23660 CZK
ABCAAB140503-100
Anti-MelanA Mouse Monoclonal Antibody [clone: 3E2]
Type: Primary
Antigen: MelanA
Clonality: monoclonal
Clone: 300
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human