Tisk…
Anti-CCR1 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB140756-50EA 25740 CZK
ABCAAB140756-50
Anti-CCR1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CCR1 (chemokine (C-C motif) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human