Tisk…

Anti-GPR83 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB140767-50EA 25590 CZK
ABCAAB140767-50
Anti-GPR83 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GPR83 (G protein-coupled receptor 83)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: extracellular domain
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR