Tisk…
Anti-LTB4R Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB140832-50EA 25740 CZK
ABCAAB140832-50
Anti-LTB4R Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LTB4R (leukotriene B4 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat