Tisk…

Anti-S1PR5 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB140932-50EA 25740 CZK
ABCAAB140932-50
Anti-S1PR5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: S1PR5 (sphingosine-1-phosphate receptor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR