Tisk…

Anti-Carbonic Anhydrase III Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB150037-100EA 24940 CZK
ABCAAB150037-100
Anti-Carbonic Anhydrase III Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Carbonic Anhydrase III
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR