Tisk…

Anti-MICA Rabbit Monoclonal Antibody

Dodavatel: Abcam
ABCAAB150355-100EA 23170 CZK
ABCAAB150355-100
Anti-MICA Rabbit Monoclonal Antibody
Type: Primary
Antigen: MICA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR