Tisk…

Anti-TAS1R3 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB150525-50EA 25650 CZK
ABCAAB150525-50
Anti-TAS1R3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TAS1R3 (taste receptor, type 1, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR