Tisk…

Anti-TM9SF3 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB150587-50EA 24420 CZK
ABCAAB150587-50
Anti-TM9SF3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TM9SF3 (transmembrane 9 superfamily member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR