Tisk…

Anti-CENPF Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB5-50EA 27580 CZK
ABCAAB5-50
Anti-CENPF Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: CENPF (centromere protein F, 350/400kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR