Tisk…

Anti-IgG Donkey Polyclonal Antibody (CF™ 594)

Dodavatel: Biotium
20116-1EA 3890 CZK
20116-1 20116.
Anti-IgG Donkey Polyclonal Antibody (CF™ 594)
Protilátky
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR