Tisk…

Anti-IgG Goat Polyclonal Antibody (CF™ 680R)

Dodavatel: Biotium
20192-1EA 3890 CZK
20192-1 20192.
Anti-IgG Goat Polyclonal Antibody (CF™ 680R)
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR