Tisk…

Anti-IgG Rabbit Polyclonal Antibody (CF™ 770)

Dodavatel: Biotium
20244-1EA 3620 CZK
20244-1 20244.
Anti-IgG Rabbit Polyclonal Antibody (CF™ 770)
Protilátky
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Guinea pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR