Tisk…
Anti-IgG2a Goat Polyclonal Antibody (CF™ 568)
Dodavatel: Biotium
20258-1EA 4380 CZK
20258-1 20258.
Anti-IgG2a Goat Polyclonal Antibody (CF™ 568)
Protilátky
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse