Tisk…

Anti-IgG2b Goat Polyclonal Antibody (CF™ 770)

Dodavatel: Biotium
20274.EA 10930 CZK
20274. 20274-1
Anti-IgG2b Goat Polyclonal Antibody (CF™ 770)
Protilátky
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR