Tisk…

Anti-HSP70 Mouse Monoclonal Antibody [clone: SJ-70]

Dodavatel: Boster
BSBTMA1050EA 15510 CZK
BSBTMA1050
Anti-HSP70 Mouse Monoclonal Antibody [clone: SJ-70]
Protilátky
Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Rabbit, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR