Tisk…

Anti-Hsp40 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA1043EA 15510 CZK
BSBTPA1043
Anti-Hsp40 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for DnaJ homolog subfamily B member 1 (DNAJB1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Hsp40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR