Tisk…

Anti-Cytokeratin 8 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA1240EA 15510 CZK
BSBTPA1240
Anti-Cytokeratin 8 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Keratin, type II cytoskeletal 8 (KRT8) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Cytokeratin 8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR