Tisk…

Anti-TGF alpha Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA1360EA 15510 CZK
BSBTPA1360
Anti-TGF alpha Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Protransforming growth factor alpha(TGFA) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: TGF alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR