Tisk…

Anti-survivin Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA1474EA 15510 CZK
BSBTPA1474
Anti-survivin Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Baculoviral IAP repeat-containing protein 5(BIRC5) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: survivin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR