Tisk…

Anti-Annexin A3 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA1510EA 15510 CZK
BSBTPA1510
Anti-Annexin A3 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Annexin A3(ANXA3) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Annexin A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR