Tisk…

Anti-Indoleamine 2, 3-dioxygenase Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA1611EA 15510 CZK
BSBTPA1611
Anti-Indoleamine 2, 3-dioxygenase Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Indoleamine 2,3-dioxygenase 1(IDO1) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Indoleamine 2, 3-dioxygenase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR