Tisk…

Anti-E2F2 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA1810EA 15510 CZK
BSBTPA1810
Anti-E2F2 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Transcription factor E2F2 (E2F2) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: E2F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR