Tisk…

Anti-Bone Sialoprotein Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA1887EA 15510 CZK
BSBTPA1887
Anti-Bone Sialoprotein Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Bone sialoprotein 2 (IBSP) detection. Tested with WB, IHC-P in Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Bone Sialoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR