Tisk…

Anti-CXCR5 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA2021EA 15510 CZK
BSBTPA2021
Anti-CXCR5 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for C-X-C chemokine receptor type 5 (CXCR5) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CXCR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR