Tisk…

Anti-CD134/OX40 Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA2136-2EA 15510 CZK
BSBTPA2136-2
Anti-CD134/OX40 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 4(TNFRSF4) detection. Tested with WB in Mouse.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CD134/OX40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR