Tisk…

Anti-Mannose 6 Phosphate Receptor (Cation independent) Rabbit Polyclonal Antibody

Dodavatel: Boster
BSBTPA2182EA 15510 CZK
BSBTPA2182
Anti-Mannose 6 Phosphate Receptor (Cation independent) Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Cation-independent mannose-6-phosphate receptor(IGF2R) detection. Tested with WB, IHC-P, ICC in Human, Mouse, Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Mannose 6 Phosphate Receptor (Cation independent)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR