Tisk…

Anti-OGG1 Rabbit Polyclonal Antibody (DyLight® 405)

Dodavatel: Novus Biologicals
NOVUNB100-106VEA 22270 CZK
NOVUNB100-106V
Anti-OGG1 Rabbit Polyclonal Antibody (DyLight® 405)
Protilátky
Type: Primary
Antigen: OGG1 (8-oxoguanine DNA glycosylase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR