Tisk…

Anti-NTHL1 Rabbit Polyclonal Antibody (DyLight® 488)

Dodavatel: Novus Biologicals
NOVUNB100-108GEA 18300 CZK
NOVUNB100-108G
Anti-NTHL1 Rabbit Polyclonal Antibody (DyLight® 488)
Protilátky
Type: Primary
Antigen: NTHL1 (nth endonuclease III-like 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR