Tisk…

Anti-DNA Polymerase epsilon Mouse Monoclonal Antibody [clone: 3C5.1]

Dodavatel: Novus Biologicals
NOVUNB100-115EA 16890 CZK
NOVUNB100-115 NOVUNB100-115SS
Anti-DNA Polymerase epsilon Mouse Monoclonal Antibody [clone: 3C5.1]
Protilátky
Type: Primary
Antigen: DNA Polymerase epsilon
Clonality: monoclonal
Clone: 3C5.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR