Tisk…

Anti-HIF1A Mouse Monoclonal Antibody HiLyte Fluor® 488 [clone: H1alpha67]

Dodavatel: Novus Biologicals
NOVUNB100-123LEA 20470 CZK
NOVUNB100-123L
Anti-HIF1A Mouse Monoclonal Antibody HiLyte Fluor® 488 [clone: H1alpha67]
Protilátky
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: H1alpha67
Conjugation: HiLyte Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR