Tisk…

Anti-CFC1 Rabbit Polyclonal Antibody (DyLight® 650)

Dodavatel: Novus Biologicals
NOVUNB100-1598CEA 20020 CZK
NOVUNB100-1598C
Anti-CFC1 Rabbit Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Primary
Antigen: CFC1 (cripto, FRL-1, cryptic family 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR